missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(propyldithio)furan 97.0+%, TCI America™

$53.90 - $236.80

Chemical Identifiers

CAS 61197-09-9
Molecular Formula C8H12OS2
Molecular Weight (g/mol) 188.303
MDL Number MFCD01729816
InChI Key YFPPCUTVJGGSQC-UHFFFAOYSA-N
Synonym 2-Methyl-3-furyl Propyl Disulfide
PubChem CID 43579
IUPAC Name 2-methyl-3-(propyldisulfanyl)furan
SMILES CCCSSC1=C(OC=C1)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M18401G
View Documents
TCI America
M18401G
1 g
Each for $53.90
Only null left
 
M18405G
View Documents
TCI America
M18405G
5 g
Each for $236.80
Only null left
 
Description

Description

Specifications

Chemical Identifiers

61197-09-9
188.303
YFPPCUTVJGGSQC-UHFFFAOYSA-N
43579
CCCSSC1=C(OC=C1)C
C8H12OS2
MFCD01729816
2-Methyl-3-furyl Propyl Disulfide
2-methyl-3-(propyldisulfanyl)furan

Specifications

61197-09-9
MFCD01729816
2810
YFPPCUTVJGGSQC-UHFFFAOYSA-N
2-methyl-3-(propyldisulfanyl)furan
43579
≥97.0% (GC)
2-Methyl-3-(propyldithio)furan
C8H12OS2
1 g
2-Methyl-3-furyl Propyl Disulfide
CCCSSC1=C(OC=C1)C
188.303
188.30
Liquid
Videos
Safety and Handling

Safety and Handling

RTECSNumber : JO1975500

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents