accessibility menu, dialog, popup

UserName

H3CSSOH3C

CAS RN 61197-09-9

IUPAC Name: 2-methyl-3-(propyldisulfanyl)furan
Synonyms: 2-methyl-3-(propyldisulfanyl)furan
Molecular Weight (g/mol): 188.3
Molecular Formula: C8H12OS2
InChi Key: YFPPCUTVJGGSQC-UHFFFAOYSA-N
SMILES: CCCSSC1=C(C)OC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results