missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(propyldithio)furan 97.0+%, TCI America™

Catalog No. M18405G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18405G 5 g
M18401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M18405G Supplier TCI America Supplier No. M18405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 61197-09-9
Molecular Formula C8H12OS2
Molecular Weight (g/mol) 188.303
MDL Number MFCD01729816
InChI Key YFPPCUTVJGGSQC-UHFFFAOYSA-N
Synonym 2-Methyl-3-furyl Propyl Disulfide
PubChem CID 43579
IUPAC Name 2-methyl-3-(propyldisulfanyl)furan
SMILES CCCSSC1=C(OC=C1)C

Specifications

Quantity 5 g
UN Number 2810
Formula Weight 188.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-(propyldithio)furan
RTECSNumber JO1975500
TSCA No
Recommended Storage Refrigerator