missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-phenyl-2-propen-1-ol 95.0+%, TCI America™

Catalog No. m05855g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M05855G Supplier TCI America Supplier No. M05855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1504-55-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00004738
InChI Key LLNAMUJRIZIXHF-VQHVLOKHSA-N
Synonym 2-methyl-3-phenyl-2-propen-1-ol, beta-methylcinnamyl alcohol, 2-propen-1-ol, 2-methyl-3-phenyl, alpha-methylcinnamyl alcohol, methyl cinnamic alcohol, alpha-methylcinnamic alcohol, alpha methyl cinnamic alcohol, 3-phenyl-2-methyl-propen-2-ol-1, e-2-methyl-3-phenyl-2-propen-1-ol, 2e-2-methyl-3-phenylprop-2-en-1-ol
PubChem CID 6393846
IUPAC Name (E)-2-methyl-3-phenylprop-2-en-1-ol
SMILES CC(=CC1=CC=CC=C1)CO

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-phenyl-2-propen-1-ol
RTECSNumber GE2340000
TSCA Yes