missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-pentanone 98.0+%, TCI America™
$70.84 - $231.14
Chemical Identifiers
| CAS | 565-69-5 |
|---|---|
| Molecular Formula | C6H12O |
| Molecular Weight (g/mol) | 100.161 |
| MDL Number | MFCD00009314 |
| InChI Key | HYTRYEXINDDXJK-UHFFFAOYSA-N |
| Synonym | 2-methyl-3-pentanone, ethyl isopropyl ketone, 3-pentanone, 2-methyl, 4-methyl-3-pentanone, isopropyl ethyl ketone, 2-methyl-3-pentanal, iso-c3h7coc2h5, acmc-209lte, ksc272m0l |
| PubChem CID | 11265 |
| IUPAC Name | 2-methylpentan-3-one |
| SMILES | CCC(=O)C(C)C |
Chemical Identifiers
| 565-69-5 | |
| 100.161 | |
| HYTRYEXINDDXJK-UHFFFAOYSA-N | |
| 11265 | |
| CCC(=O)C(C)C |
| C6H12O | |
| MFCD00009314 | |
| 2-methyl-3-pentanone, ethyl isopropyl ketone, 3-pentanone, 2-methyl, 4-methyl-3-pentanone, isopropyl ethyl ketone, 2-methyl-3-pentanal, iso-c3h7coc2h5, acmc-209lte, ksc272m0l | |
| 2-methylpentan-3-one |
Specifications
| 565-69-5 | |
| 115°C | |
| MFCD00009314 | |
| 1224 | |
| HYTRYEXINDDXJK-UHFFFAOYSA-N | |
| 2-methylpentan-3-one | |
| 11265 | |
| ≥98.0% (GC) | |
| 2-Methyl-3-pentanone |
| Yellow | |
| C6H12O | |
| 5 mL | |
| 2-methyl-3-pentanone, ethyl isopropyl ketone, 3-pentanone, 2-methyl, 4-methyl-3-pentanone, isopropyl ethyl ketone, 2-methyl-3-pentanal, iso-c3h7coc2h5, acmc-209lte, ksc272m0l | |
| CCC(=O)C(C)C | |
| 100.161 | |
| 100.16 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-2475
TSCA : No