CAS RN 565-69-5
IUPAC Name:
2-methylpentan-3-one
Synonyms:
4-methyl-3-pentanone iso-C3H7COC2H5
Molecular Weight (g/mol):
100.16
Molecular Formula:
C6H12O
InChi Key:
HYTRYEXINDDXJK-UHFFFAOYSA-N
SMILES:
CCC(=O)C(C)C
1
–
1
of
1
results
1
2-Methyl-3-pentanone 98.0+%, TCI America™
CAS: 565-69-5 Molecular Formula: C6H12O Molecular Weight (g/mol): 100.161 MDL Number: MFCD00009314 InChI Key: HYTRYEXINDDXJK-UHFFFAOYSA-N Synonym: 2-methyl-3-pentanone,ethyl isopropyl ketone,3-pentanone, 2-methyl,4-methyl-3-pentanone,isopropyl ethyl ketone,2-methyl-3-pentanal,iso-c3h7coc2h5,acmc-209lte,ksc272m0l PubChem CID: 11265 IUPAC Name: 2-methylpentan-3-one SMILES: CCC(=O)C(C)C