missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrobenzyl Chloride 98.0+%, TCI America™

Catalog No. M18165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M18165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M18165G Supplier TCI America Supplier No. M18165G
Only null left

Chemical Identifiers

CAS 60468-54-4
Molecular Formula C8H8ClNO2
Molecular Weight (g/mol) 185.61
MDL Number MFCD00007165
InChI Key XZNDXQGZPOZITR-UHFFFAOYSA-N
Synonym 2-methyl-3-nitrobenzyl chloride, 1-chloromethyl-2-methyl-3-nitrobenzene, alpha-chloro-2-methyl-3-nitrotoluene, 1-chloromethyl-2-methyl-3-nitro-benzene, 2-methyl-3-nitrobenzylchloride, benzene, 1-chloromethyl-2-methyl-3-nitro, acmc-1b2j5, ksc497o8t, 2-chloromethyl-6-nitrotoluene
PubChem CID 108947
IUPAC Name 1-(chloromethyl)-2-methyl-3-nitrobenzene
SMILES CC1=C(CCl)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 45°C
Color Yellow
Quantity 5 g
UN Number 3261
Formula Weight 185.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-3-nitrobenzyl Chloride