missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-nitroaniline 98.0+%, TCI America™
$199.46 - $1437.19
Chemical Identifiers
| CAS | 603-83-8 |
|---|---|
| Molecular Formula | C7H8N2O2 |
| Molecular Weight (g/mol) | 152.15 |
| MDL Number | MFCD00007731 |
| InChI Key | HFCFJYRLBAANKN-UHFFFAOYSA-N |
| Synonym | 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine |
| PubChem CID | 11783 |
| IUPAC Name | 2-methyl-3-nitroaniline |
| SMILES | CC1=C(N)C=CC=C1[N+]([O-])=O |
Chemical Identifiers
| 603-83-8 | |
| 152.15 | |
| HFCFJYRLBAANKN-UHFFFAOYSA-N | |
| 11783 | |
| CC1=C(N)C=CC=C1[N+]([O-])=O |
| C7H8N2O2 | |
| MFCD00007731 | |
| 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine | |
| 2-methyl-3-nitroaniline |
Specifications
| 603-83-8 | |
| 305°C | |
| MFCD00007731 | |
| 2660 | |
| HFCFJYRLBAANKN-UHFFFAOYSA-N | |
| 2-methyl-3-nitroaniline | |
| 11783 | |
| ≥98.0% (GC) | |
| 2-Methyl-3-nitroaniline |
| 90°C | |
| C7H8N2O2 | |
| 25 g | |
| 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine | |
| CC1=C(N)C=CC=C1[N+]([O-])=O | |
| 152.15 | |
| 152.15 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XU8200000
TSCA : No