missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitroaniline, 97%

CAS: 603-83-8 | C7H8N2O2 | 152.15 g/mol

$104.08 - $104.08

Chemical Identifiers

CAS 603-83-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007731
InChI Key HFCFJYRLBAANKN-UHFFFAOYSA-N
Synonym 2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine
PubChem CID 11783
IUPAC Name 2-methyl-3-nitroaniline
SMILES CC1=C(N)C=CC=C1[N+]([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Qty  
AC152020250
SDS View Documents
Thermo Scientific Chemicals
152020250
25 g Glass bottle
Each for $104.08
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

603-83-8
152.15
HFCFJYRLBAANKN-UHFFFAOYSA-N
11783
CC1=C(N)C=CC=C1[N+]([O-])=O
C7H8N2O2
MFCD00007731
2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine
2-methyl-3-nitroaniline

Specifications

603-83-8
100.0
Green-Yellow to Yellow
Authentic
Glass bottle
CH3C6H3(NO2)NH2
25 g
2-amino-6-nitrotoluene, 3-nitro-o-toluidine, benzenamine, 2-methyl-3-nitro, o-toluidine, 3-nitro, 2-methyl-3-nitro-aniline, 1-amino-2-methyl-3-nitrobenzene, 2-nitro-6-aminotoluene, 2-methyl-3-nitrobenzenamine, 2-methyl-3-nitro-phenylamine, 2-methyl-3-nitrophenyl amine
HFCFJYRLBAANKN-UHFFFAOYSA-N
2-methyl-3-nitroaniline
11783
97%
2-Methyl-3-nitroaniline
96.0
87°C to 91°C
305°C
96% min. (HPLC)
C7H8N2O2
MFCD00007731
12, 848
Solubility in water: insoluble. Other solubilities: freely soluble in ethanol and ether
CC1=C(N)C=CC=C1[N+]([O-])=O
152.15
152.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-059-6

SDS
Documents

Documents

RUO – Research Use Only