missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-hexanol, TCI America™
$82.02 - $229.29
Chemical Identifiers
| CAS | 617-29-8 |
|---|---|
| Molecular Formula | C7H16O |
| Molecular Weight (g/mol) | 116.204 |
| MDL Number | MFCD00004577 |
| InChI Key | RGRUUTLDBCWYBL-UHFFFAOYSA-N |
| PubChem CID | 12040 |
| IUPAC Name | 2-methylhexan-3-ol |
| SMILES | CCCC(C(C)C)O |
Chemical Identifiers
| 617-29-8 | |
| 116.204 | |
| RGRUUTLDBCWYBL-UHFFFAOYSA-N | |
| 2-methylhexan-3-ol |
| C7H16O | |
| MFCD00004577 | |
| 12040 | |
| CCCC(C(C)C)O |
Specifications
| 617-29-8 | |
| 145°C | |
| MFCD00004577 | |
| 1987 | |
| CCCC(C(C)C)O | |
| 116.204 | |
| 116.20 | |
| 2-Methyl-3-hexanol |
| Colorless | |
| C7H16O | |
| 5 mL | |
| RGRUUTLDBCWYBL-UHFFFAOYSA-N | |
| 2-methylhexan-3-ol | |
| 12040 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No