missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 2-Methyl-3-hexanol | |
| Colorless | |
| C7H16O | |
| 25 mL | |
| RGRUUTLDBCWYBL-UHFFFAOYSA-N | |
| 2-methylhexan-3-ol | |
| 12040 | |
| Liquid |
| 617-29-8 | |
| 145°C | |
| MFCD00004577 | |
| 1987 | |
| CCCC(C(C)C)O | |
| 116.204 | |
| 116.20 |
Chemical Identifiers
| 617-29-8 | |
| 116.204 | |
| RGRUUTLDBCWYBL-UHFFFAOYSA-N | |
| 2-methylhexan-3-ol |
| C7H16O | |
| MFCD00004577 | |
| 12040 | |
| CCCC(C(C)C)O |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No