Learn More
2-Methyl-3-butyn-2-ol, 98%
CAS: 115-19-5 | C5H8O | 84.118 g/mol
Supplier: Thermo Scientific Chemicals L07593AE
Description
2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
| 2-Methyl-3-butyn-2-ol | |
| 2°C to 3°C | |
| 103°C to 104°C | |
| C5H8O | |
| MFCD00004467 | |
| UN1987 | |
| 14,6034 | |
| Fully miscible. | |
| CC(C)(C#C)O | |
| 84.118 | |
| 84.12 |
| 115-19-5 | |
| 0.863 | |
| 22°C (72°F) | |
| 1.421 | |
| 100 mL | |
| 635746 | |
| 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol | |
| CEBKHWWANWSNTI-UHFFFAOYSA-N | |
| 2-methylbut-3-yn-2-ol | |
| 8258 | |
| 98% |
Chemical Identifiers
| 115-19-5 | |
| 84.118 | |
| CEBKHWWANWSNTI-UHFFFAOYSA-N | |
| 8258 | |
| CC(C)(C#C)O |
| C5H8O | |
| MFCD00004467 | |
| 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol | |
| 2-methylbut-3-yn-2-ol |
Safety and Handling
GHS H Statement
H225-H318-H302
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c
H225-H302-H318-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber : 204-070-5
RTECSNumber : ES0810000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only