missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-butyn-2-ol, 98%

Catalog No. p-7044647
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL07593AE 100 mL
AAL07593AP 500 mL
AAL075930F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAL07593AE Supplier Thermo Scientific Chemicals Supplier No. L07593AE
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 115-19-5 | C5H8O | 84.118 g/mol

2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3-butyn-2-ol is used as an intermediate in the manufacture of products for the agrochemical and specialty chemical industry.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 115-19-5
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00004467
InChI Key CEBKHWWANWSNTI-UHFFFAOYSA-N
Synonym 2-methyl-3-butyn-2-ol, 3-butyn-2-ol, 2-methyl, 3-methyl-1-butyn-3-ol, 3-methylbutynol, 2-methyl-2-butynol, ethynyldimethylcarbinol, carbavane, dimethylethynylcarbinol, dimethylethynylmethanol, 1,1-dimethyl-2-propynol
PubChem CID 8258
IUPAC Name 2-methylbut-3-yn-2-ol
SMILES CC(C)(C#C)O

Specifications

Melting Point 2°C to 3°C
Density 0.863
Boiling Point 103°C to 104°C
Flash Point 22°C (72°F)
Refractive Index 1.421
Quantity 100 mL
UN Number UN1987
Beilstein 635746
Merck Index 14,6034
Solubility Information Fully miscible.
Formula Weight 84.12
Percent Purity 98%
Chemical Name or Material 2-Methyl-3-butyn-2-ol
Show More Show Less
Hazard Category H225-H302-H318-H336
Hazard Statement GHS H Statement
H225-H318-H302
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 204-070-5
RTECSNumber ES0810000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only