missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-2-propenyl)succinic Anhydride 97.0+%, TCI America™

Catalog No. M112325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M112325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M112325G Supplier TCI America Supplier No. M112325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18908-20-8
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00075477
InChI Key ANPMHDUGFOMMJJ-UHFFFAOYNA-N
Synonym (beta-Methallyl)succinic Anhydride, 4-Methyl-4-pentene-1,2-dicarboxylic Anhydride
PubChem CID 225375
IUPAC Name 3-(2-methylprop-2-en-1-yl)oxolane-2,5-dione
SMILES CC(=C)CC1CC(=O)OC1=O

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Boiling Point 137°C
Formula Weight 154.17
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material (2-Methyl-2-propenyl)succinic Anhydride