accessibility menu, dialog, popup

UserName

CH3H2COOO

CAS RN 18908-20-8

IUPAC Name: 3-(2-methylprop-2-en-1-yl)oxolane-2,5-dione
Synonyms: 3-(2-methylprop-2-en-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: ANPMHDUGFOMMJJ-UHFFFAOYNA-N
SMILES: CC(=C)CC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results