missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propenyl Acetate 98.0+%, TCI America™

Catalog No. A160525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A160525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A160525ML Supplier TCI America Supplier No. A160525ML
Only null left

Chemical Identifiers

CAS 820-71-3
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00048215
InChI Key IVKYUXHYUAMPMT-UHFFFAOYSA-N
Synonym methallyl acetate, 2-methylallyl acetate, 2-methylprop-2-en-1-yl acetate, 2-propen-1-ol, 2-methyl-, acetate, 2-methyl-2-propenyl acetate, beta-methallyl acetate, acmc-209pmw, .beta.-methallyl acetate, 1-acetoxy-2-methyl-2-propene, acetic acid, 2-methylallylester
PubChem CID 69962
IUPAC Name 2-methylprop-2-enyl acetate
SMILES CC(=C)COC(=O)C

Specifications

Color Yellow
Quantity 25 mL
UN Number 3272
Formula Weight 114.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-propenyl Acetate