CAS RN 820-71-3
CAS RN 820-71-3
IUPAC Name:
2-methylprop-2-en-1-yl acetate
Synonyms:
2-methylprop-2-en-1-yl acetate
Molecular Weight (g/mol):
114.14
Molecular Formula:
C6H10O2
InChi Key:
IVKYUXHYUAMPMT-UHFFFAOYSA-N
SMILES:
CC(=C)COC(C)=O
1
–
1
of
1
results
1
2-Methyl-2-propenyl Acetate 98.0+%, TCI America™
CAS: 820-71-3 Molecular Formula: C6H10O2 Molecular Weight (g/mol): 114.144 MDL Number: MFCD00048215 InChI Key: IVKYUXHYUAMPMT-UHFFFAOYSA-N Synonym: methallyl acetate,2-methylallyl acetate,2-methylprop-2-en-1-yl acetate,2-propen-1-ol, 2-methyl-, acetate,2-methyl-2-propenyl acetate,beta-methallyl acetate,acmc-209pmw,.beta.-methallyl acetate,1-acetoxy-2-methyl-2-propene,acetic acid, 2-methylallylester PubChem CID: 69962 IUPAC Name: 2-methylprop-2-enyl acetate SMILES: CC(=C)COC(=O)C