missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-phenylpropanenitrile, 97%

Catalog No. p-4228820
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438250010 1 g
AC438250050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438250010 Supplier Thermo Scientific Chemicals Supplier No. 438250010

CAS: 1195-98-8 | C10H11N | 145.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1195-98-8
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.21
InChI Key PGQTYXFMSZUGOW-UHFFFAOYSA-N
Synonym 2-methyl-2-phenyl-propionitrile, phenylisobutyronitrile, 2-phenylisobutyronitrile, 2-methyl-2-phenylpropionitrile, 2-methyl-2-phenyl-propanenitrile, alpha,alpha-dimethylbenzyl cyanide, benzeneacetonitrile,a,a-dimethyl, alpha,alpha-dimethylphenylacetonitrile, benzeneacetonitrile,, a,, a-dimethyl, benzeneacetonitrile, alpha,alpha-dimethyl
PubChem CID 2797591
IUPAC Name 2-methyl-2-phenylpropanenitrile
SMILES CC(C)(C#N)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 54°C to 56°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 145.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-phenylpropanenitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only