missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentenal, (E)+(Z), 97%

Catalog No. p-7022808
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1572914 25 g
AAA1572922 100 g
AAA1572936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1572914 Supplier Thermo Scientific Chemicals Supplier No. A1572914
Only null left

CAS: 623-36-9 | C6H10O | 98.145 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 623-36-9
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00006978
InChI Key IDEYZABHVQLHAF-GQCTYLIASA-N
Synonym 2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
PubChem CID 5319754
IUPAC Name (E)-2-methylpent-2-enal
SMILES CCC=C(C)C=O

Specifications

Melting Point -90°C
Density 0.86
Boiling Point 134°C to 136°C
Flash Point 31°C (87°F)
Odor Pleasant
Refractive Index 1.4495
Quantity 25 g
UN Number UN1988
Beilstein 506124
Sensitivity Air Sensitive
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-2-pentenal, (E)+(Z)
Show More Show Less
Hazard Category H226-H317-H318-H331-H335
Hazard Statement GHS H Statement
H331-H318-H226-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber 210-789-5
RTECSNumber SB2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only