accessibility menu, dialog, popup

UserName

CAS RN 623-36-9

IUPAC Name: (2E)-2-methylpent-2-enal
Synonyms: 2-pentenal, 2-methyl-
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: IDEYZABHVQLHAF-GQCTYLIASA-N
SMILES: CC\C=C(/C)C=O
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)