Learn More
2-Methyl-2-pentenal, 97%
CAS: 623-36-9 | C6H10O | 98.15 g/mol
$76.57 - $76.57
Chemical Identifiers
| CAS | 623-36-9 |
|---|---|
| Molecular Formula | C6H10O |
| Molecular Weight (g/mol) | 98.15 |
| MDL Number | MFCD00006978 |
| InChI Key | IDEYZABHVQLHAF-GQCTYLIASA-N |
| Synonym | 2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530 |
| PubChem CID | 5319754 |
| IUPAC Name | (E)-2-methylpent-2-enal |
| SMILES | CCC=C(C)C=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC359960250
|
Thermo Scientific Chemicals
359960250 |
25 g | Glass bottle |
Each for $76.57
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 623-36-9 | |
| 98.15 | |
| IDEYZABHVQLHAF-GQCTYLIASA-N | |
| 5319754 | |
| CCC=C(C)C=O |
| C6H10O | |
| MFCD00006978 | |
| 2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530 | |
| (E)-2-methylpent-2-enal |
Specifications
| 623-36-9 | |
| 100.0 | |
| 0.8600g/mL | |
| 31°C | |
| 96% min. (GC) | |
| C6H10O | |
| C2H5CH=C(CH3)CHO | |
| 25 g | |
| 2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530 | |
| IDEYZABHVQLHAF-GQCTYLIASA-N | |
| (E)-2-methylpent-2-enal | |
| 5319754 | |
| 97% | |
| 2-Methyl-2-pentenal |
| 96.0 | |
| Colorless to Yellow | |
| 134.0°C to 136.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4470 to 1.4550 | |
| MFCD00006978 | |
| 0.86 | |
| Solubility in water: insoluble | |
| CCC=C(C)C=O | |
| 98.15 | |
| 98.15 | |
| Liquid |
Safety and Handling
GHS H Statement
Toxic if inhaled.
May cause an allergic skin reaction.
Causes serious eye damage.
Flammable liquid and vapor.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 210-789-5
RUO – Research Use Only