missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-nitro-1,3-propanediol 98.0+%, TCI America™
Supplier: TCI America M063925G
Specifications
| 2-Methyl-2-nitro-1,3-propanediol | |
| 156°C | |
| C4H9NO4 | |
| 25 g | |
| LOTYADDQWWVBDJ-UHFFFAOYSA-N | |
| 2-methyl-2-nitropropane-1,3-diol | |
| 6480 | |
| ≥98.0% (GC) |
| 77-49-6 | |
| White-Yellow | |
| MFCD00024812 | |
| 2-methyl-2-nitro-1,3-propanediol, nmpd, 2-nitro-2-methyl-1,3-propanediol, nitroisobutylglycol, 1,3-propanediol, 2-methyl-2-nitro, 2-methyl-2-nitropropanediol, 1,1-dimethylol-1-nitroethane, caswell no. 573n, epa pesticide chemical code 573900, acmc-1basy | |
| CC(CO)(CO)[N+]([O-])=O | |
| 135.12 | |
| 135.12 | |
| Crystalline Powder |
Chemical Identifiers
| 77-49-6 | |
| 135.12 | |
| LOTYADDQWWVBDJ-UHFFFAOYSA-N | |
| 6480 | |
| CC(CO)(CO)[N+]([O-])=O |
| C4H9NO4 | |
| MFCD00024812 | |
| 2-methyl-2-nitro-1,3-propanediol, nmpd, 2-nitro-2-methyl-1,3-propanediol, nitroisobutylglycol, 1,3-propanediol, 2-methyl-2-nitro, 2-methyl-2-nitropropanediol, 1,1-dimethylol-1-nitroethane, caswell no. 573n, epa pesticide chemical code 573900, acmc-1basy | |
| 2-methyl-2-nitropropane-1,3-diol |
Safety and Handling
RTECSNumber : TY8925000
TSCA : Yes