missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-hexanol 96.0+%, TCI America™
$242.33 - $848.14
Chemical Identifiers
| CAS | 625-23-0 |
|---|---|
| Molecular Formula | C7H16O |
| Molecular Weight (g/mol) | 116.204 |
| MDL Number | MFCD00004486 |
| InChI Key | KRIMXCDMVRMCTC-UHFFFAOYSA-N |
| Synonym | 2-methyl-2-hexanol, butyldimethylcarbinol, 2-hexanol, 2-methyl, tert-heptyl alcohol, 1,1-dimethyl-1-pentanol, tert-heptanol 9ci, methyl-2 hexanol-2, 2-methyl-2-hexanoi, 1-pentanol, dimethyl, 2-methyl-2-hydroxyhexane |
| PubChem CID | 12240 |
| IUPAC Name | 2-methylhexan-2-ol |
| SMILES | CCCCC(C)(C)O |
Chemical Identifiers
| 625-23-0 | |
| 116.204 | |
| KRIMXCDMVRMCTC-UHFFFAOYSA-N | |
| 12240 | |
| CCCCC(C)(C)O |
| C7H16O | |
| MFCD00004486 | |
| 2-methyl-2-hexanol, butyldimethylcarbinol, 2-hexanol, 2-methyl, tert-heptyl alcohol, 1,1-dimethyl-1-pentanol, tert-heptanol 9ci, methyl-2 hexanol-2, 2-methyl-2-hexanoi, 1-pentanol, dimethyl, 2-methyl-2-hydroxyhexane | |
| 2-methylhexan-2-ol |
Specifications
| 625-23-0 | |
| 143°C | |
| MFCD00004486 | |
| 1987 | |
| KRIMXCDMVRMCTC-UHFFFAOYSA-N | |
| 2-methylhexan-2-ol | |
| 12240 | |
| ≥96.0% (GC) | |
| 2-Methyl-2-hexanol |
| Yellow | |
| C7H16O | |
| 5 g | |
| 2-methyl-2-hexanol, butyldimethylcarbinol, 2-hexanol, 2-methyl, tert-heptyl alcohol, 1,1-dimethyl-1-pentanol, tert-heptanol 9ci, methyl-2 hexanol-2, 2-methyl-2-hexanoi, 1-pentanol, dimethyl, 2-methyl-2-hydroxyhexane | |
| CCCCC(C)(C)O | |
| 116.204 | |
| 116.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : No