accessibility menu, dialog, popup

UserName

H3CCH3CH3HO

CAS RN 625-23-0

IUPAC Name: 2-methylhexan-2-ol
Synonyms: 2-methyl-2-hexanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: KRIMXCDMVRMCTC-UHFFFAOYSA-N
SMILES: CCCCC(C)(C)O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results