missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-hexanol 96.0+%, TCI America™

Catalog No. M05425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M05425G 5 g
M054225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M05425G Supplier TCI America Supplier No. M05425G
Only null left

Chemical Identifiers

CAS 625-23-0
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.204
MDL Number MFCD00004486
InChI Key KRIMXCDMVRMCTC-UHFFFAOYSA-N
Synonym 2-methyl-2-hexanol, butyldimethylcarbinol, 2-hexanol, 2-methyl, tert-heptyl alcohol, 1,1-dimethyl-1-pentanol, tert-heptanol 9ci, methyl-2 hexanol-2, 2-methyl-2-hexanoi, 1-pentanol, dimethyl, 2-methyl-2-hydroxyhexane
PubChem CID 12240
IUPAC Name 2-methylhexan-2-ol
SMILES CCCCC(C)(C)O

Specifications

Color Yellow
Boiling Point 143°C
Quantity 5 g
UN Number 1987
Formula Weight 116.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-hexanol
EINECSNumber (2)-0217
TSCA No