missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butenenitrile 70.0+%, TCI America™

Catalog No. p-7132394
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
M083925ML 25 mL
M0839500ML 500 mL
2 options

Catalog No. M083925ML

Supplier: TCI America M083925ML

Only null left

Chemical Identifiers

CAS 4403-61-6
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00037169
InChI Key IHXNSHZBFXGOJM-HYXAFXHYSA-N
Synonym 2-Cyano-2-butene, 1-Methyl-1-propenyl Cyanide
PubChem CID 6028739
IUPAC Name (Z)-2-methylbut-2-enenitrile
SMILES CC=C(C)C#N

Specifications

Color Yellow
Boiling Point 50°C
Quantity 25 mL
UN Number 3273
Formula Weight 81.12
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butenenitrile
RTECSNumber EM8312500
TSCA No