CAS RN 4403-61-6
CAS RN 4403-61-6
IUPAC Name:
(2E)-2-methylbut-2-enenitrile
Synonyms:
2-butenenitrile, 2-methyl-
Molecular Weight (g/mol):
81.12
Molecular Formula:
C5H7N
InChi Key:
IHXNSHZBFXGOJM-HWKANZROSA-N
SMILES:
C\C=C(/C)C#N
1
–
1
of
1
results
1
2-Methyl-2-butenenitrile 70.0+%, TCI America™
CAS: 4403-61-6 Molecular Formula: C5H7N Molecular Weight (g/mol): 81.118 MDL Number: MFCD00037169 InChI Key: IHXNSHZBFXGOJM-HYXAFXHYSA-N Synonym: 2-Cyano-2-butene, 1-Methyl-1-propenyl Cyanide PubChem CID: 6028739 IUPAC Name: (Z)-2-methylbut-2-enenitrile SMILES: CC=C(C)C#N