missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butenenitrile 70.0+%, TCI America™

Catalog No. M083925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M083925ML 25 mL
M0839500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M083925ML Supplier TCI America Supplier No. M083925ML
Only null left

Chemical Identifiers

CAS 4403-61-6
Molecular Formula C5H7N
Molecular Weight (g/mol) 81.118
MDL Number MFCD00037169
InChI Key IHXNSHZBFXGOJM-HYXAFXHYSA-N
Synonym 2-Cyano-2-butene, 1-Methyl-1-propenyl Cyanide
PubChem CID 6028739
IUPAC Name (Z)-2-methylbut-2-enenitrile
SMILES CC=C(C)C#N

Specifications

Color Yellow
Boiling Point 50°C
Quantity 25 mL
UN Number 3273
Formula Weight 81.12
Percent Purity ≥70.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butenenitrile
RTECSNumber EM8312500
TSCA No