missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene 95.0+%, TCI America™

Supplier:  TCI America P0067100ML

 View more versions of this product

Catalog No. P0067100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methyl-2-butene
-134°C
38°C
MFCD00009276
2460
BKOOMYPCSUNDGP-UHFFFAOYSA-N
2-methylbut-2-ene
10553
70.14
Liquid
513-35-9
Colorless
C5H10
100 mL
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CC=C(C)C
70.14
CHEBI:77916
≥95.0% (GC)

Chemical Identifiers

513-35-9
70.14
BKOOMYPCSUNDGP-UHFFFAOYSA-N
10553
2-methylbut-2-ene
C5H10
MFCD00009276
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CHEBI:77916
CC=C(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0019

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions