missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene 90.0+%, TCI America™

Catalog No. p-7132311
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M070825ML 25 mL
M0708100ML 100 mL
M0708500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M070825ML Supplier TCI America Supplier No. M070825ML
Only null left

Chemical Identifiers

CAS 513-35-9
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009276
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
PubChem CID 10553
ChEBI CHEBI:77916
IUPAC Name 2-methylbut-2-ene
SMILES CC=C(C)C

Specifications

Melting Point -134°C
Color Colorless
Boiling Point 38°C
Quantity 25 mL
UN Number 2460
Formula Weight 70.14
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butene
EINECSNumber (2)-0019
TSCA Yes