missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 98.0+%, TCI America™
Supplier: TCI America M2717200MG
Specifications
| 2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole | |
| 172°C | |
| C12H14N2 | |
| 200 mg | |
| CN1CCC2=C(C1)C3=CC=CC=C3N2 | |
| 186.26 | |
| 186.26 | |
| Crystalline Powder |
| 5094-12-2 | |
| White-Yellow | |
| MFCD00460783 | |
| FYHWPFXPFFPQRT-UHFFFAOYSA-N | |
| 2-methyl-1,3,4,5-tetrahydropyrido[4,3-b]indole | |
| 97424 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 5094-12-2 | |
| 186.26 | |
| FYHWPFXPFFPQRT-UHFFFAOYSA-N | |
| 2-methyl-1,3,4,5-tetrahydropyrido[4,3-b]indole |
| C12H14N2 | |
| MFCD00460783 | |
| 97424 | |
| CN1CCC2=C(C1)C3=CC=CC=C3N2 |
Safety and Handling
RTECSNumber : UV0830000
TSCA : No