missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole 98.0+%, TCI America™

Catalog No. p-7133386
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2717200MG 200 mg
M27171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2717200MG Supplier TCI America Supplier No. M2717200MG
Only null left

Chemical Identifiers

CAS 5094-12-2
Molecular Formula C12H14N2
Molecular Weight (g/mol) 186.26
MDL Number MFCD00460783
InChI Key FYHWPFXPFFPQRT-UHFFFAOYSA-N
PubChem CID 97424
IUPAC Name 2-methyl-1,3,4,5-tetrahydropyrido[4,3-b]indole
SMILES CN1CCC2=C(C1)C3=CC=CC=C3N2

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 200 mg
Formula Weight 186.26
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-2,3,4,5-tetrahydro-1H-pyrido[4,3-b]indole
RTECSNumber UV0830000
TSCA No