Learn More
2-Methyl-1-tetralone, 97%, Thermo Scientific Chemicals
$300.10 - $300.10
Chemical Identifiers
| CAS | 1590-08-5 |
|---|---|
| Molecular Formula | C11H12O |
| Molecular Weight (g/mol) | 160.216 |
| MDL Number | MFCD00001690 |
| InChI Key | GANIBVZSZGNMNB-UHFFFAOYSA-N |
| Synonym | 2-methyl-1-tetralone, 2-methyl-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-2-methyl, 2-methyltetralin-1-one, 2-methyl-1,2,3,4-tetrahydronaphthalen-1-one, 2-methyl-2,3,4-trihydronaphthalen-1-one, 2-methyltetralone, 2-methyl-1-tetralon, acmc-20anr7, 2-methyl-alpha-tetralone |
| PubChem CID | 102650 |
| IUPAC Name | 2-methyl-3,4-dihydro-2H-naphthalen-1-one |
| SMILES | CC1CCC2=CC=CC=C2C1=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3186503
|
Thermo Scientific Chemicals
H3186503 |
1 g |
Each for $300.10
|
|
|||||
Description
Methylation of 1-tetralone followed by the subsequent dehydrogenation of 2-methyl-1-tetralone in the presence of Pd/C catalyst also has been reported.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethylation of 1-tetralone followed by the subsequent dehydrogenation of 2-methyl-1-tetralone in the presence of Pd/C catalyst also has been reported.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 1590-08-5 | |
| 160.216 | |
| GANIBVZSZGNMNB-UHFFFAOYSA-N | |
| 102650 | |
| CC1CCC2=CC=CC=C2C1=O |
| C11H12O | |
| MFCD00001690 | |
| 2-methyl-1-tetralone, 2-methyl-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-2-methyl, 2-methyltetralin-1-one, 2-methyl-1,2,3,4-tetrahydronaphthalen-1-one, 2-methyl-2,3,4-trihydronaphthalen-1-one, 2-methyltetralone, 2-methyl-1-tetralon, acmc-20anr7, 2-methyl-alpha-tetralone | |
| 2-methyl-3,4-dihydro-2H-naphthalen-1-one |
Specifications
| 1590-08-5 | |
| 129°C (12 mmHg) | |
| C11H12O | |
| MFCD00001690 | |
| 2-methyl-1-tetralone, 2-methyl-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-2-methyl, 2-methyltetralin-1-one, 2-methyl-1,2,3,4-tetrahydronaphthalen-1-one, 2-methyl-2,3,4-trihydronaphthalen-1-one, 2-methyltetralone, 2-methyl-1-tetralon, acmc-20anr7, 2-methyl-alpha-tetralone | |
| GANIBVZSZGNMNB-UHFFFAOYSA-N | |
| 2-methyl-3,4-dihydro-2H-naphthalen-1-one | |
| 102650 | |
| 98% |
| 1.058 | |
| >110°C (230°F) | |
| 1.552 | |
| 1 g | |
| Insoluble in water. | |
| CC1CCC2=CC=CC=C2C1=O | |
| 160.216 | |
| 160.22 | |
| 2-Methyl-1-tetralone |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 216-461-8
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only