missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol 99.0+%, TCI America™

Catalog No. p-7131208
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
I009425ML 25 mL
I0094500ML 500 mL
2 options

Catalog No. I009425ML

Supplier: TCI America I009425ML

Only null left

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 1212
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-propanol
EINECSNumber (2)-3049
RTECSNumber NP9625000
TSCA Yes