missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol 99.0+%, TCI America™

Catalog No. I009425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I009425ML 25 mL
I0094500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I009425ML Supplier TCI America Supplier No. I009425ML

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Boiling Point 108°C
Quantity 25 mL
UN Number 1212
Formula Weight 74.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-propanol
EINECSNumber (2)-3049
RTECSNumber NP9625000
TSCA Yes