missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-Propanol, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™

Catalog No. p-7120070
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046181 1 L Poly Bottle
60046182 2.5 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046181 Supplier Honeywell Chemicals Supplier No. 330641L
Only null left

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Color Colorless
Density 0.803g/mL at 25°C
pH 7 (at 80g/L)
Boiling Point 108°C
Flash Point 28°C (82.4°F)
Ignition Point 801°F
Packaging Poly Bottle
Refractive Index n20/D 1.396
Linear Formula (CH3)2CHCH2OH
Quantity 1 L
UN Number UN1212
Beilstein 1730878
Solubility Information In water 70g/L at 20°C
Vapor Density 2.55 (vs air)
Vapor Pressure 8mmHg (20°C), 8.8mmHg (0°C)
Formula Weight 74.12g/mol
Percent Purity ≥99% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material 2-Methyl-1-Propanol, Reag. Ph. Eur.
Show More Show Less
Hazard Statement P210-P280-P304 + P340 + P312-P305 + P351 + P338 + P310-P403 + P235
Precautionary Statement H226-H315-H318-H335-H336
DOTInformation Class 3 - PG 3 - Isobutanol
EINECSNumber 201-148-0
RTECSNumber NP9625000