Learn More
2-Methyl-1-propanol, 99+%, Extra Dry, AcroSeal™
CAS: 78-83-1 | C4H10O | 74.12 g/mol
Supplier: Thermo Scientific Chemicals 398950010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methyl-1-propanol | |
| 78-83-1 | |
| 100.0 | |
| 0.8000g/mL | |
| 28°C | |
| Glass Bottle | |
| 1.3945 to 1.3965 | |
| MFCD00004740 | |
| 01, 373 | |
| 15, 5176 | |
| Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone | |
| CC(C)CO | |
| 74.12 | |
| 4 Pas (20°C) | |
| 74.12 | |
| Extra Dry |
| Extra Dry | |
| 99.0 | |
| -108.0°C | |
| 108.0°C | |
| 99% min. (GC) | |
| C4H10O | |
| (CH3)2CHCH2OH | |
| 1 L | |
| 0.8 | |
| 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 2-methylpropan-1-ol | |
| 6560 | |
| CHEBI:46645 | |
| 99+% |
Chemical Identifiers
| 78-83-1 | |
| 74.12 | |
| ZXEKIIBDNHEJCQ-UHFFFAOYSA-N | |
| 6560 | |
| 2-methylpropan-1-ol |
| C4H10O | |
| MFCD00004740 | |
| 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol | |
| CHEBI:46645 | |
| CC(C)CO |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
GHS Signal Word: Danger
EINECSNumber : 201-148-
RUO – Research Use Only