missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-phenylpropene 98.0+%, TCI America™
$251.65 - $708.33
Chemical Identifiers
| CAS | 768-49-0 |
|---|---|
| Molecular Formula | C10H12 |
| Molecular Weight (g/mol) | 132.206 |
| MDL Number | MFCD00008899 |
| InChI Key | BTOVVHWKPVSLBI-UHFFFAOYSA-N |
| Synonym | (2-Methylpropenyl)benzene |
| PubChem CID | 13030 |
| IUPAC Name | 2-methylprop-1-enylbenzene |
| SMILES | CC(=CC1=CC=CC=C1)C |
Chemical Identifiers
| 768-49-0 | |
| 132.206 | |
| BTOVVHWKPVSLBI-UHFFFAOYSA-N | |
| 13030 | |
| CC(=CC1=CC=CC=C1)C |
| C10H12 | |
| MFCD00008899 | |
| (2-Methylpropenyl)benzene | |
| 2-methylprop-1-enylbenzene |
Specifications
| 768-49-0 | |
| 189°C | |
| MFCD00008899 | |
| 3295 | |
| BTOVVHWKPVSLBI-UHFFFAOYSA-N | |
| 2-methylprop-1-enylbenzene | |
| 13030 | |
| ≥98.0% (GC) | |
| 2-Methyl-1-phenylpropene |
| Colorless | |
| C10H12 | |
| 1 g | |
| (2-Methylpropenyl)benzene | |
| CC(=CC1=CC=CC=C1)C | |
| 132.206 | |
| 132.21 | |
| Liquid |
Safety and Handling
TSCA : Yes