missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenylpropene 98.0+%, TCI America™

Supplier:  TCI America M10151G

 View more versions of this product

Catalog No. M10151G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methyl-1-phenylpropene
Colorless
C10H12
1 g
(2-Methylpropenyl)benzene
CC(=CC1=CC=CC=C1)C
132.206
132.21
Liquid
768-49-0
189°C
MFCD00008899
3295
BTOVVHWKPVSLBI-UHFFFAOYSA-N
2-methylprop-1-enylbenzene
13030
≥98.0% (GC)

Chemical Identifiers

768-49-0
132.206
BTOVVHWKPVSLBI-UHFFFAOYSA-N
13030
CC(=CC1=CC=CC=C1)C
C10H12
MFCD00008899
(2-Methylpropenyl)benzene
2-methylprop-1-enylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions