missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-2-propen-1-ol, tech. 85%

Catalog No. p-7025458
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2093606 5 g
AAB2093614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2093606 Supplier Thermo Scientific Chemicals Supplier No. B2093606
Only null left

CAS: 4383-08-8 | C10H12O | 148.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4383-08-8
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.21
MDL Number MFCD00046630
InChI Key ZGYBYYJGIKPBFD-UHFFFAOYNA-N
Synonym 2-methyl-1-phenyl-2-propen-1-ol, acmc-1bn9z, alpha-isopropenylbenzenemethanol, 1-phenyl-2-methyl-2-propen-1-ol, 2,6-dichloro-9-methyl-10h-purine
PubChem CID 138211
IUPAC Name 2-methyl-1-phenylprop-2-en-1-ol
SMILES CC(=C)C(C1=CC=CC=C1)O

Specifications

Quantity 5 g
Formula Weight 148.21
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Methyl-1-phenyl-2-propen-1-ol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only