missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-2-propanol 98.0+%, TCI America™

Catalog No. D078325ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D078325ML 25 mL
D0783100ML 100 mL
D0783500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D078325ML Supplier TCI America Supplier No. D078325ML

Chemical Identifiers

CAS 100-86-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00004465
InChI Key RIWRBSMFKVOJMN-UHFFFAOYSA-N
Synonym 2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinol
PubChem CID 7531
IUPAC Name 2-methyl-1-phenylpropan-2-ol
SMILES CC(C)(CC1=CC=CC=C1)O

Specifications

Color White-Yellow
Boiling Point 94°C
Quantity 25 mL
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methyl-1-phenyl-2-propanol
EINECSNumber (3)-1034
RTECSNumber SG8050000
TSCA Yes