Learn More
2-Methyl-1-phenyl-2-propanol, 98+%
CAS: 100-86-7 | C10H14O | 150.221 g/mol
Supplier: Thermo Scientific Chemicals L0367922
Description
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2-Methyl-1-phenyl-2-propanol | |
| 23°C to 25°C | |
| 215°C | |
| C10H14O | |
| MFCD00004465 | |
| 1855608 | |
| Slightly soluble in water. | |
| CC(C)(CC1=CC=CC=C1)O | |
| 150.221 | |
| 150.22 |
| 100-86-7 | |
| 0.974 | |
| 82°C (179°F) | |
| 1.515 | |
| 100 g | |
| 2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinol | |
| RIWRBSMFKVOJMN-UHFFFAOYSA-N | |
| 2-methyl-1-phenylpropan-2-ol | |
| 7531 | |
| ≥98% |
Chemical Identifiers
| 100-86-7 | |
| 150.221 | |
| RIWRBSMFKVOJMN-UHFFFAOYSA-N | |
| 7531 | |
| CC(C)(CC1=CC=CC=C1)O |
| C10H14O | |
| MFCD00004465 | |
| 2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinol | |
| 2-methyl-1-phenylpropan-2-ol |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 202-896-0
RTECSNumber : SG8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only