missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-2-propanol, 98+%

Catalog No. AAL0367936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0367936 500 g
AAL0367922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0367936 Supplier Thermo Scientific Chemicals Supplier No. L0367936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 100-86-7 | C10H14O | 150.221 g/mol

2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 100-86-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00004465
InChI Key RIWRBSMFKVOJMN-UHFFFAOYSA-N
Synonym 2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinol
PubChem CID 7531
IUPAC Name 2-methyl-1-phenylpropan-2-ol
SMILES CC(C)(CC1=CC=CC=C1)O

Specifications

Melting Point 23°C to 25°C
Density 0.974
Boiling Point 215°C
Flash Point 82°C (179°F)
Refractive Index 1.515
Quantity 500 g
Beilstein 1855608
Solubility Information Slightly soluble in water.
Formula Weight 150.22
Percent Purity ≥98%
Chemical Name or Material 2-Methyl-1-phenyl-2-propanol
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-896-0
RTECSNumber SG8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only