missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-1-propanol, 98%

Catalog No. aab2072906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2072906 5 g
AAB2072914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2072906 Supplier Thermo Scientific Chemicals Supplier No. B2072906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 611-69-8 | C10H14O | 150.221 g/mol

It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker's yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker′s yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 611-69-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00065000
InChI Key GMDYDZMQHRTHJA-UHFFFAOYSA-N
Synonym 2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9
PubChem CID 95626
IUPAC Name 2-methyl-1-phenylpropan-1-ol
SMILES CC(C)C(C1=CC=CC=C1)O

Specifications

Density 0.964
Boiling Point 124°C to 125°C (15 mmHg)
Flash Point 86°C (187°F)
Refractive Index 1.513
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material 2-Methyl-1-phenyl-1-propanol
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 210-274-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only