missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pentene 98.0+%, TCI America™

Supplier:  TCI America M0245100ML

 View more versions of this product

Catalog No. M0245100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Methyl-1-pentene
-136°C
61°C
MFCD00009405
2288
WWUVJRULCWHUSA-UHFFFAOYSA-N
2-methylpent-1-ene
12986
≥98.0% (GC)
763-29-1
Colorless
C6H12
100 mL
2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
CCCC(=C)C
84.162
84.16
Liquid

Chemical Identifiers

763-29-1
84.162
WWUVJRULCWHUSA-UHFFFAOYSA-N
12986
CCCC(=C)C
C6H12
MFCD00009405
2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
2-methylpent-1-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0031

RTECSNumber : SB2230000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions