missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pentene 98.0+%, TCI America™

Catalog No. p-7132075
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M024525ML 25 mL
M0245100ML 100 mL
M0245500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M024525ML Supplier TCI America Supplier No. M024525ML
Only null left

Chemical Identifiers

CAS 763-29-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009405
InChI Key WWUVJRULCWHUSA-UHFFFAOYSA-N
Synonym 2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
PubChem CID 12986
IUPAC Name 2-methylpent-1-ene
SMILES CCCC(=C)C

Specifications

Melting Point -136°C
Color Colorless
Boiling Point 61°C
Quantity 25 mL
UN Number 2288
Formula Weight 84.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-pentene
EINECSNumber (2)-0031
RTECSNumber SB2230000
TSCA Yes