missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-pentanol 98.0+%, TCI America™
$100.67 - $721.40
Chemical Identifiers
| CAS | 105-30-6 |
|---|---|
| Molecular Formula | C6H14O |
| Molecular Weight (g/mol) | 102.177 |
| MDL Number | MFCD00004745 |
| InChI Key | PFNHSEQQEPMLNI-UHFFFAOYSA-N |
| Synonym | 2-methyl-1-pentanol, 1-pentanol, 2-methyl, 2-methylpentanol-1, 2-methylpentanol, sec-amyl carbinol, 2-methylamyl alcohol, +--2-methylpentanol, 2-mpoh, +--2-methyl-1-pentanol, 2-methyl-2-propylethanol |
| PubChem CID | 7745 |
| IUPAC Name | 2-methylpentan-1-ol |
| SMILES | CCCC(C)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M019225ML
|
TCI America
M019225ML |
25 mL |
Each for $100.67
|
|
|||||
|
M0192500ML
|
TCI America
M0192500ML |
500 mL |
Each for $721.40
|
|
|||||
Chemical Identifiers
| 105-30-6 | |
| 102.177 | |
| PFNHSEQQEPMLNI-UHFFFAOYSA-N | |
| 7745 | |
| CCCC(C)CO |
| C6H14O | |
| MFCD00004745 | |
| 2-methyl-1-pentanol, 1-pentanol, 2-methyl, 2-methylpentanol-1, 2-methylpentanol, sec-amyl carbinol, 2-methylamyl alcohol, +--2-methylpentanol, 2-mpoh, +--2-methyl-1-pentanol, 2-methyl-2-propylethanol | |
| 2-methylpentan-1-ol |
Specifications
| 105-30-6 | |
| 148°C | |
| MFCD00004745 | |
| 1987 | |
| PFNHSEQQEPMLNI-UHFFFAOYSA-N | |
| 2-methylpentan-1-ol | |
| 7745 | |
| ≥98.0% (GC) | |
| 2-Methyl-1-pentanol |
| Colorless | |
| C6H14O | |
| 25 mL | |
| 2-methyl-1-pentanol, 1-pentanol, 2-methyl, 2-methylpentanol-1, 2-methylpentanol, sec-amyl carbinol, 2-methylamyl alcohol, +--2-methylpentanol, 2-mpoh, +--2-methyl-1-pentanol, 2-methyl-2-propylethanol | |
| CCCC(C)CO | |
| 102.177 | |
| 102.18 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : SA7175000
TSCA : Yes