missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-nitronaphthalene 98.0+%, TCI America™

Catalog No. M037625G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M037625G 25 g
M0376250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M037625G Supplier TCI America Supplier No. M037625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 881-03-8
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.20
MDL Number MFCD00003914
InChI Key IZNWACYOILBFEG-UHFFFAOYSA-N
Synonym 1-nitro-2-methylnaphthalene, naphthalene, 2-methyl-1-nitro, 2-methyl-1-nitrophthalene, unii-0ig2r1r9f7, 2-methyl-1-nitro-naphthalene, ccris 4680, methylnitronaphthalene, pubchem11253, dsstox_cid_5761, acmc-209qr4
PubChem CID 13432
IUPAC Name 2-methyl-1-nitronaphthalene
SMILES CC1=CC=C2C=CC=CC2=C1[N+]([O-])=O

Specifications

Melting Point 81°C
Boiling Point 188°C
Quantity 25 g
Formula Weight 187.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1-nitronaphthalene