accessibility menu, dialog, popup

UserName

H3CCH3H2C

CAS RN 563-46-2

IUPAC Name: 2-methylbut-1-ene
Synonyms: methylbutenes
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: MHNNAWXXUZQSNM-UHFFFAOYSA-N
SMILES: CCC(C)=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)