missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-butene 98.0+%, TCI America™

Catalog No. M017425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M017425ML 25 mL
M0174100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M017425ML Supplier TCI America Supplier No. M017425ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 563-46-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009333
InChI Key MHNNAWXXUZQSNM-UHFFFAOYSA-N
Synonym 2-methyl-1-butene, 2-methylbutene, 1-butene, 2-methyl, 1-isoamylene, 2-methylbutene-1, butene, 2-methyl, gamma-isoamylene, .gamma.-isoamylene, unii-33c9y0i55h, c2h5c ch3 =ch2
PubChem CID 11240
ChEBI CHEBI:77915
IUPAC Name 2-methylbut-1-ene
SMILES CCC(C)=C

Specifications

Color Colorless
Boiling Point 31°C
Quantity 25 mL
UN Number 2459
Formula Weight 70.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-butene
EINECSNumber (2)-0019
TSCA Yes
Recommended Storage Refrigerator